Tetrathiafulvalene ≥99.0%, purified by sublimation
Leverantör: TCI EUROPE
Synonyms:
2,2'-Bi(1,3-dithiolylidene), TTF
T3380-200MG
T3380-1G
TCIAT3380-200MGEA
485
SEK
TCIAT3380-200MG
TCIAT3380-1G
Tetrathiafulvalene ≥99.0%, purified by sublimation
Tetrathiafulvalene
Formel:
C₆H₄S₄ MW: 204,356 g/mol |
MDL Number:
MFCD00005492 CAS nummer: 31366-25-3 EINECS: 250-593-7 Merck Index: 14,09242 |
Specifikation testresultat
Appearance | Orange to Brown powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 118.0 to 123.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...