2,4,6-Tribromopyridine ≥97.0%
Leverantör: TCI
|
Varning
|
T3375-1G
T3375-5G
TCIAT3375-1GEA
1325
SEK
TCIAT3375-1G
TCIAT3375-5G
2,4,6-Tribromopyridine ≥97.0%
2,4,6-Tribromopyridine
Formel:
C₅H₂Br₃N Kokpunkt: 284°C |
MDL Number:
MFCD00234002 CAS nummer: 2408-70-0 |
|
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 107.0 to 111.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...