o-Tolyl trifluoromethanesulphonate ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Trifluoromethanesulphonic acid o-Tolyl ester, o-tolyl Trifluoromethanesulphonate, o-Tolyl triflate

T3357-5G T3357-25G
TCIAT3357-5GEA 1375 SEK
TCIAT3357-5G TCIAT3357-25G
o-Tolyl trifluoromethanesulphonate ≥98.0%
o-Tolyl trifluoromethanesulphonate
Formel: C₈H₇F₃O₃S
MW: 240,20 g/mol
MDL Number: MFCD29089375
CAS nummer: 66107-34-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=630f0fd04a838c48142f6177712b4431b90ebe0b&vid=630f0fd04a838c48142f6177712b4431b90ebe0b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR