9,3':6',9''-Tercarbazole ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI
Varning

T3344-200MG T3344-1G
TCIAT3344-200MGEA 645 SEK
TCIAT3344-200MG TCIAT3344-1G
9,3':6',9''-Tercarbazole ≥98.0% (by HPLC, total nitrogen)
9,3':6',9''-Tercarbazole
Formel: C₃₆H₂₃N₃
Smältpunkt: 285°C
MDL Number: MFCD29472452
CAS nummer: 606129-90-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f248a80258a26a355a09b16576df167b22b6f4f3&vid=f248a80258a26a355a09b16576df167b22b6f4f3&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR