4-(Trifluoromethoxy)phenyl isothiocyanate ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 4-(Trifluoromethoxy)phenylisothiocyanate

T3341-1G T3341-5G
TCIAT3341-1GEA 390 SEK
TCIAT3341-1G TCIAT3341-5G
4-(Trifluoromethoxy)phenyl isothiocyanate ≥98.0%
4-(Trifluoromethoxy)phenyl isothiocyanate
Formel: C₈H₄F₃NOS
MW: 219,19 g/mol
Kokpunkt: 88°C
Flampunkt: 107°C
Storage Temperature: Kylskåp
MDL Number: MFCD00042412
CAS nummer: 64285-95-6
UN: 1760
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53417afaec7f6626c07e36fcbe24f249561edc20&vid=53417afaec7f6626c07e36fcbe24f249561edc20&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR