9,9',10,10'-Tetraphenyl-2,2'-bianthracene ≥97.0%

Leverantör: TCI EUROPE

T3336-200MG T3336-1G
TCIAT3336-200MGEA 1225 SEK
TCIAT3336-200MG TCIAT3336-1G
9,9',10,10'-Tetraphenyl-2,2'-bianthracene ≥97.0%
9,9',10,10'-Tetraphenyl-2,2'-bianthracene
Formel: C₅₂H₃₄
MW: 658,84 g/mol
CAS nummer: 172285-72-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5dd0843c4310c119759e39e3a51a3f64a02a3ed&vid=d5dd0843c4310c119759e39e3a51a3f64a02a3ed&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Green powder to crystal
Purity (HPLC) min. 97.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR