1,2,3-Tribromobenzene ≥98.0%

Leverantör: TCI EUROPE
Varning

T3329-1G T3329-5G
TCIAT3329-1GEA 1250 SEK
TCIAT3329-1G TCIAT3329-5G
1,2,3-Tribromobenzene ≥98.0%
1,2,3-Tribromobenzene
Formel: C₆H₃Br₃
MW: 314,80 g/mol
Kokpunkt: 283°C
MDL Number: MFCD00192668
CAS nummer: 608-21-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b5fc06a45e44a89ecce121d17e00e37b4ea5252&vid=6b5fc06a45e44a89ecce121d17e00e37b4ea5252&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 86.0 to 90.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR