2,4,6-Trimethylphenyl isothiocyanate ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 1-Isothiocyanato-2,4,6-trimethylbenzene, 2,4,6-Trimethylphenylisothiocyanate

T3324-5G T3324-25G
TCIAT3324-5GEA 1300 SEK
TCIAT3324-5G TCIAT3324-25G
2,4,6-Trimethylphenyl isothiocyanate ≥98.0%
2,4,6-Trimethylphenyl isothiocyanate
Formel: C₁₀H₁₁NS
MW: 177,27 g/mol
Kokpunkt: 75°C
MDL Number: MFCD00022055
CAS nummer: 6095-82-5
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89dacad7aea8dbc6d8e51d235efc2e5ee651be17&vid=89dacad7aea8dbc6d8e51d235efc2e5ee651be17&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 61.0 to 65.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR