4,4,5,5-Tetramethyl-2-(triphenylen-2-yl)-1,3,2-dioxaborolane ≥97.0% (by HPLC, total nitrogen)
Leverantör: TCI
Synonyms:
Triphenylen-2-ylboronic acid Pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)triphenylene
T3322-1G
T3322-5G
TCIAT3322-1GEA
1400
SEK
TCIAT3322-1G
TCIAT3322-5G
4,4,5,5-Tetramethyl-2-(triphenylen-2-yl)-1,3,2-dioxaborolane ≥97.0% (by HPLC, total nitrogen)
4,4,5,5-Tetramethyl-2-(triphenylen-2-yl)-1,3,2-dioxaborolane
Formel:
C₂₄H₂₃BO₂ MW: 354.26 g/mol |
MDL Number:
MFCD26401844 CAS nummer: 890042-13-4 |
|
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 174.0 to 178.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...