2',3,4,5-Tetrafluoro-4'-(trans-4-propylcyclohexyl)biphenyl ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
rel-2,3',4',5'-Tetrafluoro-4-((1s,4r)-4-propylcyclohexyl)-1,1'-biphenyl
T3317-1G
T3317-5G
TCIAT3317-1GEA
375
SEK
TCIAT3317-1G
TCIAT3317-5G
2',3,4,5-Tetrafluoro-4'-(trans-4-propylcyclohexyl)biphenyl ≥98.0%
2',3,4,5-Tetrafluoro-4'-(trans-4-propylcyclohexyl)biphenyl
Formel:
C₂₁H₂₂F₄ MW: 350,40 g/mol |
MDL Number:
MFCD22380673 CAS nummer: 173837-35-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 61.0 to 65.0 deg-C |
Mesomorphic range | 61.0 to 66.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...