2-Triphenylenecarboxaldehyde ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
Triphenylene-2-carbaldehyde
T3304-200MG
T3304-1G
TCIAT3304-200MGEA
1450
SEK
TCIAT3304-200MG
TCIAT3304-1G
2-Triphenylenecarboxaldehyde ≥98.0%
2-Triphenylenecarboxaldehyde
Formel:
C₁₉H₁₂O MW: 256,30 g/mol Smältpunkt: 162°C |
MDL Number:
MFCD29089374 CAS nummer: 96404-79-4 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...