Teriflunomide ≥98.0%

Leverantör: TCI
Varning

Synonyms: N-[4-(Trifluoromethyl)phenyl]-2-cyano-3-hydroxycrotonamide, (Z)-2-Cyano-3-hydroxy-N-[4-(trifluoromethyl)phenyl]-2-butenamide, (Z)-2-Cyano-3-hydroxy-N-(4-(trifluoromethyl)phenyl)but-2-enamide

T3287-1G T3287-100MG
TCIAT3287-1GEA 3975 SEK
TCIAT3287-1G TCIAT3287-100MG
Teriflunomide ≥98.0%
Teriflunomide
Formel: C₁₂H₉F₃N₂O₂
Storage Temperature: Frys
MDL Number: MFCD00910058
CAS nummer: 163451-81-8
Merck Index: 14,09165
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c4df2e3ecda99cd5a50cced7723af08bcda9637&vid=2c4df2e3ecda99cd5a50cced7723af08bcda9637&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 227.0 to 231.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR