p-Terphenyl ≥99.5%, purified by sublimation
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-Phenyl-1,1'-biphenyl, 1,4-Diphenylbenzene, 4-Phenylbiphenyl, 1,4-Terphenyl
T3263-5G
T3263-25G
TCIAT3263-5GEA
430
SEK
TCIAT3263-5G
TCIAT3263-25G
p-Terphenyl ≥99.5%, purified by sublimation
p-Terphenyl
Formel:
C₁₈H₁₄ MW: 230,31 g/mol Kokpunkt: 389°C |
MDL Number:
MFCD00003061 CAS nummer: 92-94-4 EINECS: 202-205-2 |
Specifikation testresultat
Appearance | White powder to crystal |
Purity (GC) | min. 99.5 % |
Melting point | 212.0 to 216.0 deg-C |
Solubility in hot Toluene | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...