3-(Perfluorohexyl)propanol-1 ≥98.0%

Leverantör: TCI
Varning

T3258-5G T3258-25G
TCIAT3258-5GEA 870 SEK
TCIAT3258-5G TCIAT3258-25G
3-(Perfluorohexyl)propanol-1 ≥98.0%
3-(Perfluorohexyl)propanol-1
Formel: C₉H₇F₁₃O
Kokpunkt: 86°C
Smältpunkt: -3°C
MDL Number: MFCD00077578
CAS nummer: 80806-68-4
UN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac9403c25f2e2d198260dea16e9140af42a9c929&vid=ac9403c25f2e2d198260dea16e9140af42a9c929&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR