1-Methoxy-2,2,2-trifluoroethanol ≥88.0% (contains ca. 10% Methanol)
Leverantör: TCI EUROPE
Fara
Synonyms: Trifluoroacetaldehyde methyl hemiacetal
T3247-5G T3247-25G
TCIAT3247-5GEA 470 SEK
TCIAT3247-5G TCIAT3247-25G
1-Methoxy-2,2,2-trifluoroethanol ≥88.0% (contains ca. 10% Methanol)
1-Methoxy-2,2,2-trifluoroethanol
Formel: C₃H₅F₃O₂
MW: 130,07 g/mol
Flampunkt: 39°C
MDL Number: MFCD00013572
CAS nummer: 431-46-9
EINECS: 207-072-4
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7378e7967ba220262fc1d8e2e57f0d3d7ada77c4&vid=7378e7967ba220262fc1d8e2e57f0d3d7ada77c4&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 88.0 %
Methanol max. 10.0 %