Triethoxy[5,5,6,6,7,7,7-heptafluoro-4,4-bis(trifluoromethyl)heptyl]silane ≥90.0%

Leverantör: TCI
Varning

T3246-1G T3246-5G
TCIAT3246-1GEA 1025 SEK
TCIAT3246-1G TCIAT3246-5G
Triethoxy[5,5,6,6,7,7,7-heptafluoro-4,4-bis(trifluoromethyl)heptyl]silane ≥90.0%
Triethoxy[5,5,6,6,7,7,7-heptafluoro-4,4-bis(trifluoromethyl)heptyl]silane
Formel: C₁₅H₂₁F₁₃O₃Si
MW: 524.39 g/mol
Kokpunkt: 81°C
MDL Number: MFCD27918526
CAS nummer: 130676-81-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd37f5ca8b5add8b7153eb209a7c72aae485ce02&vid=dd37f5ca8b5add8b7153eb209a7c72aae485ce02&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR