α,α,α-Trifluoro-3,5-diaminotoluene ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
5-(Trifluoromethyl)-1,3-phenylenediamine, 5-(Trifluoromethyl)-m-phenylenediamine, 5-Trifluoromethyl-1,3-phenylenediamine, 5-(Trifluoromethyl)benzene-1,3-diamine, 3,5-Diaminobenzotrifluoride
T3239-1G
T3239-5G
TCIAT3239-1GEA
510
SEK
TCIAT3239-1G
TCIAT3239-5G
α,α,α-Trifluoro-3,5-diaminotoluene ≥98.0% (by GC, titration analysis)
α,α,α-Trifluoro-3,5-diaminotoluene
Formel:
C₇H₇F₃N₂ MW: 176,14 g/mol |
MDL Number:
MFCD00014776 CAS nummer: 368-53-6 EINECS: 206-708-8 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 87.0 to 91.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...