Thieno[3,2-b]thiophene-2,5-dicarboxaldehyde ≥93.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Thieno[3,2-b]thiophene-2,5-dicarbaldehyde
T3212-200MG
T3212-1G
TCIAT3212-200MGEA
655
SEK
TCIAT3212-200MG
TCIAT3212-1G
Thieno[3,2-b]thiophene-2,5-dicarboxaldehyde ≥93.0%
Thieno[3,2-b]thiophene-2,5-dicarboxaldehyde
Formel:
C₈H₄O₂S₂ MW: 196,24 g/mol Smältpunkt: 274°C |
MDL Number:
MFCD03931117 CAS nummer: 37882-75-0 |
Specifikation testresultat
Appearance | Orange to Brown powder to crystal |
Purity (GC) | min. 93.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...