Trimethoxy(4-vinylphenyl)silane ≥97.0%
Leverantör: TCI EUROPE
Varning
T3185-5G T3185-25G
TCIAT3185-5GEA 1100 SEK
TCIAT3185-5G TCIAT3185-25G
Trimethoxy(4-vinylphenyl)silane ≥97.0%
Trimethoxy(4-vinylphenyl)silane
Formel: C₁₁H₁₆O₃Si
Kokpunkt: 77°C
MDL Number: MFCD21596662
CAS nummer: 18001-13-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2fefa74a2e8baa1f6b10c9849004ef769fa954e&vid=d2fefa74a2e8baa1f6b10c9849004ef769fa954e&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %