Triptycene-9,10-dicarboxaldehyde ≥96.0%
Leverantör: TCI EUROPE
Synonyms:
9,10-Diformyltriptycene
T3146-200MG
T3146-1G
TCIAT3146-200MGEA
1875
SEK
TCIAT3146-200MG
TCIAT3146-1G
Triptycene-9,10-dicarboxaldehyde ≥96.0%
Triptycene-9,10-dicarboxaldehyde
Formel:
C₂₂H₁₄O₂ |
CAS nummer:
467429-73-8 |
Specifikation testresultat
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...