Tetrakis(4-hydroxyphenyl)ethylene ≥97.0%

Leverantör: TCI

T3142-1G T3142-5G
TCIAT3142-1GEA 2775 SEK
TCIAT3142-1G TCIAT3142-5G
Tetrakis(4-hydroxyphenyl)ethylene ≥97.0%
Tetrakis(4-hydroxyphenyl)ethylene
Formel: C₂₆H₂₀O₄
MW: 396.44 g/mol
Smältpunkt: 323°C
MDL Number: MFCD28901376
CAS nummer: 119301-59-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3390b4e774c310fdbb317f9f0ecb4429915a4b9c&vid=3390b4e774c310fdbb317f9f0ecb4429915a4b9c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR