Tris(1H-benzotriazol-1-yl)methane ≥95.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Tris(1H-benzo[d][1,2,3]triazol-1-yl)methane

T3141-200MG T3141-1G
TCIAT3141-200MGEA 1050 SEK
TCIAT3141-200MG TCIAT3141-1G
Tris(1H-benzotriazol-1-yl)methane ≥95.0%
Tris(1H-benzotriazol-1-yl)methane
Formel: C₁₉H₁₃N₉
MW: 367,38 g/mol
Smältpunkt: 191°C
MDL Number: MFCD00514776
CAS nummer: 88088-95-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=097aeab37f2e3cbc51424fc7d5329f6b88f90fa6&vid=097aeab37f2e3cbc51424fc7d5329f6b88f90fa6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR