N,N,2-Trimethyl-5-nitrobenzenesulphonamide ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
Synonyms:
BRL-50481, BRL 50481
T3112-200MG
T3112-1G
TCIAT3112-200MGEA
1175
SEK
TCIAT3112-200MG
TCIAT3112-1G
N,N,2-Trimethyl-5-nitrobenzenesulphonamide ≥98.0% (by HPLC, total nitrogen)
N,N,2-Trimethyl-5-nitrobenzenesulphonamide
Formel:
C₉H₁₂N₂O₄S MW: 244,27 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD03039912 CAS nummer: 433695-36-4 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 71.0 to 75.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...