Tazarotene ≥98.0%
Leverantör: TCI
Synonyms:
Ethyl 6-((4,4-dimethylthiochroman-6-yl)ethynyl)nicotinate
T3108-10MG
T3108-100MG
TCIAT3108-10MGEA
375
SEK
TCIAT3108-10MG
TCIAT3108-100MG
Tazarotene ≥98.0%
Tazarotene
Formel:
C₂₁H₂₁NO₂S Storage Temperature: Kylskåp |
MDL Number:
MFCD00867628 CAS nummer: 118292-40-3 Merck Index: 14,09081 |
|
Specifikation testresultat
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 102.0 to 106.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...