Tripalmitin (glycerol tripalmitate) ≥80.0%
Leverantör: TCI EUROPE
Synonyms:
Propane-1,2,3-triyl tripalmitate, Tripalmitin, Glycerol tripalmitate
T3096-25G
T3096-100G
TCIAT3096-25GEA
630
SEK
TCIAT3096-25G
TCIAT3096-100G
Tripalmitin (glycerol tripalmitate) ≥80.0%
Tripalmitin (glycerol tripalmitate)
Formel:
C₅₁H₉₈O₆ MW: 807,34 g/mol Kokpunkt: 320°C |
MDL Number:
MFCD00008995 CAS nummer: 555-44-2 EINECS: 209-098-1 Merck Index: 14,09736 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 80.0 % |
Melting point | 63.0 to 68.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...