1,3,5-Tris(4-iodophenyl)benzene ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4,4''-Diiodo-5'-(4-iodophenyl)-1,1':3',1''-terphenyl
T3084-1G
T3084-5G
TCIAT3084-1GEA
1750
SEK
TCIAT3084-1G
TCIAT3084-5G
1,3,5-Tris(4-iodophenyl)benzene ≥98.0%
1,3,5-Tris(4-iodophenyl)benzene
Formel:
C₂₄H₁₅I₃ MW: 684,10 g/mol Smältpunkt: 266°C |
MDL Number:
MFCD07369795 CAS nummer: 151417-38-8 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...