4-(Trifluoromethyl)piperidine ≥97.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-Trifluoromethylpiperidine
T3081-1G
T3081-5G
TCIAT3081-1GEA
1075
SEK
TCIAT3081-1G
TCIAT3081-5G
4-(Trifluoromethyl)piperidine ≥97.0% (by GC, titration analysis)
4-(Trifluoromethyl)piperidine
Formel:
C₆H₁₀F₃N MW: 153,15 g/mol Kokpunkt: 135°C |
MDL Number:
MFCD00102068 CAS nummer: 657-36-3 UN: 1993 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...