1-(4-(Trifluoromethyl)benzyl)piperazine ≥97.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 1-(4-Trifluoromethylbenzyl)piperazine

T3075-1G T3075-5G
TCIAT3075-1GEA 645 SEK
TCIAT3075-1G TCIAT3075-5G
1-(4-(Trifluoromethyl)benzyl)piperazine ≥97.0% (by GC, titration analysis)
1-(4-(Trifluoromethyl)benzyl)piperazine
Formel: C₁₂H₁₅F₃N₂
MW: 244,26 g/mol
Kokpunkt: 100°C
MDL Number: MFCD03407490
CAS nummer: 107890-32-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e69df65f46e4373069ab9b0b317c8fb30cab5055&vid=e69df65f46e4373069ab9b0b317c8fb30cab5055&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR