3-(2-Thienyl)pyridine ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3-(Thiophen-2-yl)pyridine

T3055-200MG T3055-1G
TCIAT3055-200MGEA 830 SEK
TCIAT3055-200MG TCIAT3055-1G
3-(2-Thienyl)pyridine ≥98.0%
3-(2-Thienyl)pyridine
Formel: C₉H₇NS
MW: 161,23 g/mol
Kokpunkt: 100°C
MDL Number: MFCD01860052
CAS nummer: 21298-53-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74fd2c800231ad57328c6f4eabf8fc2564297aee&vid=74fd2c800231ad57328c6f4eabf8fc2564297aee&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR