1,3,6,8-Tetraphenylpyrene ≥98.0%

Leverantör: TCI

T3042-50MG T3042-200MG
TCIAT3042-50MGEA 1425 SEK
TCIAT3042-50MG TCIAT3042-200MG
1,3,6,8-Tetraphenylpyrene ≥98.0%
1,3,6,8-Tetraphenylpyrene
Formel: C₄₀H₂₆
MW: 506.65 g/mol
MDL Number: MFCD00021662
CAS nummer: 13638-82-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b333136cf9f16028c7d26ab417c39b00732f276b&vid=b333136cf9f16028c7d26ab417c39b00732f276b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR