Tris(pyrazol-1-yl)methane ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Tris(1-pyrazolyl)methane

T3026-1G T3026-5G
TCIAT3026-1GEA 1675 SEK
TCIAT3026-1G TCIAT3026-5G
Tris(pyrazol-1-yl)methane ≥98.0%
Tris(pyrazol-1-yl)methane
Formel: C₁₀H₁₀N₆
MW: 214,23 g/mol
MDL Number: MFCD00040255
CAS nummer: 80510-03-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd29204bea48593250faa5d4857720c32086d12d&vid=dd29204bea48593250faa5d4857720c32086d12d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystaline
Purity (GC) min. 98.0 %
Melting point 102.0 to 106.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR