(Pentafluoroethyl)trimethylsilane ≥98.0%

Leverantör: TCI EUROPE
Fara

T3011-1G T3011-5G
TCIAT3011-1GEA 895 SEK
TCIAT3011-1G TCIAT3011-5G
(Pentafluoroethyl)trimethylsilane ≥98.0%
(Pentafluoroethyl)trimethylsilane
Formel: C₅H₉F₅Si
MW: 192,21 g/mol
Kokpunkt: 70°C
MDL Number: MFCD04116436
CAS nummer: 124898-13-1
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb103556830a2662b284f0b564e450ef4d545b9e&vid=fb103556830a2662b284f0b564e450ef4d545b9e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR