2,2,4,4-Tetramethyl-3-pentanone imine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: Di-tert-butylmethanimine, 2,2,4,4-Tetramethyl-3-pentanimine
T2989-1G T2989-5G
TCIAT2989-1GEA 1225 SEK
TCIAT2989-1G TCIAT2989-5G
2,2,4,4-Tetramethyl-3-pentanone imine ≥98.0% (by GC, titration analysis)
2,2,4,4-Tetramethyl-3-pentanone imine
Formel: C₉H₁₉N
MW: 141,26 g/mol
Kokpunkt: 164°C
Flampunkt: 29°C
Storage Temperature: Kylskåp
MDL Number: MFCD00216566
CAS nummer: 29097-52-7
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=334276ba9aac1c7884ad3edefa725a31477aec73&vid=334276ba9aac1c7884ad3edefa725a31477aec73&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %