3,3,3-Trifluoro-1-propanol ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
3,3,3-Trifluoropropan-1-ol
T2986-1G
T2986-5G
TCIAT2986-1GEA
455
SEK
TCIAT2986-1G
TCIAT2986-5G
3,3,3-Trifluoro-1-propanol ≥98.0%
3,3,3-Trifluoro-1-propanol
Formel:
C₃H₅F₃O Kokpunkt: 100°C Flampunkt: 37°C |
MDL Number:
MFCD00190649 CAS nummer: 2240-88-2 UN: 1987 ADR: 3,III |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...