(E)-Trimethyl(3,3,3-trifluoro-1-propenyl)silane ≥95.0%

Leverantör: TCI
Fara

T2977-1G T2977-5G
TCIAT2977-1GEA 9650 SEK
TCIAT2977-1G TCIAT2977-5G
(E)-Trimethyl(3,3,3-trifluoro-1-propenyl)silane ≥95.0%
(E)-Trimethyl(3,3,3-trifluoro-1-propenyl)silane
Formel: C₆H₁₁F₃Si
Kokpunkt: 69°C
CAS nummer: 55364-28-8
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce8a31dbf04082fefc6a8a8e8bb78bc479a4c689&vid=ce8a31dbf04082fefc6a8a8e8bb78bc479a4c689&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR