1,3,6,8-Tetrabromocarbazole ≥95.0%

Leverantör: TCI
Varning

T2932-200MG T2932-1G
TCIAT2932-200MGEA 1025 SEK
TCIAT2932-200MG TCIAT2932-1G
1,3,6,8-Tetrabromocarbazole ≥95.0%
1,3,6,8-Tetrabromocarbazole
Formel: C₁₂H₅Br₄N
MW: 482.80 g/mol
MDL Number: MFCD00218272
CAS nummer: 55119-09-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=780aeffbd3ae7f0b76e41965c4c61ec113bfb600&vid=780aeffbd3ae7f0b76e41965c4c61ec113bfb600&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 227.0 to 231.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR