2-Thiopheneboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Synonyms:
4,4,5,5-Tetramethyl-2-(2-thienyl)-1,3,2-dioxaborolane
T2924-1G
T2924-5G
TCIAT2924-1GEA
285
SEK
TCIAT2924-1G
TCIAT2924-5G
2-Thiopheneboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2-Thiopheneboronic acid pinacol ester
Formel:
C₁₀H₁₅BO₂S MW: 210.1 g/mol |
MDL Number:
MFCD05663878 CAS nummer: 193978-23-3 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 66.0 to 70.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...