Trimethoxy(1H,1H,2H,2H-nonafluorohexyl)silane ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1H,1H,2H,2H-Nonafluorohexyltrimethoxysilane, Trimethoxy(3,3,4,4,5,5,6,6,6-nonafluorohexyl)silane, Trimethoxy(1H,1H,2H,2H-perfluorohexyl)silane
T2918-5G
T2918-25G
TCIAT2918-5GEA
1350
SEK
TCIAT2918-5G
TCIAT2918-25G
Trimethoxy(1H,1H,2H,2H-nonafluorohexyl)silane ≥97.0%
Trimethoxy(1H,1H,2H,2H-nonafluorohexyl)silane
Formel:
C₉H₁₃F₉O₃Si MW: 368,27 g/mol Kokpunkt: 43°C Flampunkt: 71°C |
MDL Number:
MFCD08275518 CAS nummer: 85877-79-8 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...