Trimethoxy(1H,1H,2H,2H-heptadecafluorodecyl)silane ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 1H,1H,2H,2H-Heptadecafluorodecyltrimethoxysilane, Trimethoxy(1H,1H,2H,2H-perfluorodecyl)silane, (3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecyl)trimethoxysilane
T2917-5G T2917-25G
TCIAT2917-5GEA 2150 SEK
TCIAT2917-5G TCIAT2917-25G
Trimethoxy(1H,1H,2H,2H-heptadecafluorodecyl)silane ≥98.0%
Trimethoxy(1H,1H,2H,2H-heptadecafluorodecyl)silane
Formel: C₁₃H₁₃F₁₇O₃Si
MW: 568,30 g/mol
Kokpunkt: 80°C
MDL Number: MFCD07368748
CAS nummer: 83048-65-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d09c67ac1fa7ba91a9cdbc46ce690aab60546bc&vid=0d09c67ac1fa7ba91a9cdbc46ce690aab60546bc&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %