Triethoxy(1H,1H,2H,2H-nonafluorohexyl)silane ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1H,1H,2H,2H-Nonafluorohexyltriethoxysilane, Triethoxy(1H,1H,2H,2H-perfluorohexyl)silane
T2860-25G
T2860-5G
TCIAT2860-25GEA
3250
SEK
TCIAT2860-25G
TCIAT2860-5G
Triethoxy(1H,1H,2H,2H-nonafluorohexyl)silane ≥97.0%
Triethoxy(1H,1H,2H,2H-nonafluorohexyl)silane
Formel:
C₁₂H₁₉F₉O₃Si MW: 410,35 g/mol |
MDL Number:
MFCD09954454 CAS nummer: 102390-98-7 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...