N,N,N',N'-Tetrakis(2-hydroxyethyl)adipamide ≥98.0%

Leverantör: TCI

Synonyms: N,N,N',N'-Tetrakis(2-hydroxyethyl)hexanediamide, N1,N1,N6,N6-Tetrakis(2-hydroxyethyl)adipamide

T2859-5G T2859-25G
TCIAT2859-5GEA 725 SEK
TCIAT2859-5G TCIAT2859-25G
N,N,N',N'-Tetrakis(2-hydroxyethyl)adipamide ≥98.0%
N,N,N',N'-Tetrakis(2-hydroxyethyl)adipamide
Formel: C₁₄H₂₈N₂O₆
MW: 320.39 g/mol
MDL Number: MFCD07787618
CAS nummer: 6334-25-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cba38f490f837228d25ac26e9a4c3c6034496a95&vid=cba38f490f837228d25ac26e9a4c3c6034496a95&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 126.0 to 130.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR