BAPTA tetrasodium salt 98,0 Titrimetrisk

Leverantör: TCI

Synonyms: 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid tetrasodium Salt, Tetrasodium-1,2-bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetate, BAPTA-Na4

T2844-200MG T2844-1G
TCIAT2844-200MGEA 550 SEK
TCIAT2844-200MG TCIAT2844-1G
BAPTA tetrasodium salt 98,0 Titrimetrisk
BAPTA tetrasodium salt
Formel: C₂₂H₂₄N₂O₁₀
MW: 564.37 g/mol
Storage Temperature: Frys
MDL Number: MFCD00067691
CAS nummer: 126824-24-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a14b3f7aea0f58af8441c62cda2909687f1dbf0&vid=3a14b3f7aea0f58af8441c62cda2909687f1dbf0&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity (HPLC) min. 95.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Water max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR