Triethyl(trifluoromethyl)silane ≥98.0%
Leverantör: TCI
Fara
Synonyms: (Triethylsilyl)trifluoromethane
T2764-1G T2764-5G
TCIAT2764-1GEA 845 SEK
TCIAT2764-1G TCIAT2764-5G
Triethyl(trifluoromethyl)silane ≥98.0%
Triethyl(trifluoromethyl)silane
Formel: C₇H₁₅F₃Si
MW: 184.28 g/mol
Kokpunkt: 58°C
Flampunkt: 0°C
MDL Number: MFCD00192520
CAS nummer: 120120-26-5
UN: 1993
ADR: 3 II
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %