Temozolomide ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 3,4-Dihydro-3-methyl-4-oxoimidazo[5,1-d][1,2,3,5]tetrazine-8-carboxamide, 3-Methyl-4-oxo-3,4-dihydroimidazo[5,1-d][1,2,3,5]tetrazine-8-carboxamide

T2744-5G T2744-500MG
TCIAT2744-5GEA 1925 SEK
TCIAT2744-5G TCIAT2744-500MG
Temozolomide ≥98.0%
Temozolomide
Formel: C₆H₆N₆O₂
MW: 194,15 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00866492
CAS nummer: 85622-93-1
Merck Index: 14,09139

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR