Thieno[2,3-b]thiophene ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1,6-Dithiapentalene

T2729-1G T2729-5G
TCIAT2729-1GEA 1475 SEK
TCIAT2729-1G TCIAT2729-5G
Thieno[2,3-b]thiophene ≥97.0%
Thieno[2,3-b]thiophene
Formel: C₆H₄S₂
MW: 140,22 g/mol
Kokpunkt: 226°C
Smältpunkt: 7°C
Flampunkt: 93°C
MDL Number: MFCD02824126
CAS nummer: 250-84-0

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR