3,3',5,5'-Tetrabromo-2,2'-bithiophene ≥98.0%

Leverantör: TCI EUROPE

T2699-1G T2699-25G T2699-5G
TCIAT2699-1GEA 405 SEK
TCIAT2699-1G TCIAT2699-25G TCIAT2699-5G
3,3',5,5'-Tetrabromo-2,2'-bithiophene ≥98.0%
3,3',5,5'-Tetrabromo-2,2'-bithiophene
Formel: C₈H₂Br₄S₂
MW: 481,84 g/mol
MDL Number: MFCD00114807
CAS nummer: 125143-53-5

Order Now

Specifikation testresultat

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 138.0 to 144.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR