1,2,3,4-Tetramethyl-1,2,3,4-cyclobutanetetracarboxylic dianhydride
Leverantör: TCI
|
Varning
|
T2697-1G
T2697-5G
TCIAT2697-1GEA
1325
SEK
TCIAT2697-1G
TCIAT2697-5G
1,2,3,4-Tetramethyl-1,2,3,4-cyclobutanetetracarboxylic dianhydride
1,2,3,4-Tetramethyl-1,2,3,4-cyclobutanetetracarboxylic dianhydride
Formel:
C₁₂H₁₂O₆ MW: 252.22 g/mol Smältpunkt: 380°C |
CAS nummer:
64198-16-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Elemental analysis (Carbon) | 56.00 to 57.70 % |