2',3',5'-Tri-O-acetyl-6-chloro-2-iodopurine riboside ≥97.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE

Synonyms: 6-Chloro-2-iodo-9-(2,3,5-tri-O-acetyl-beta-D-ribofuranosyl)purine

T2691-1G T2691-5G
TCIAT2691-1GEA 2525 SEK
TCIAT2691-1G TCIAT2691-5G
2',3',5'-Tri-O-acetyl-6-chloro-2-iodopurine riboside ≥97.0% (by HPLC, total nitrogen)
2',3',5'-Tri-O-acetyl-6-chloro-2-iodopurine riboside
Formel: C₁₆H₁₆ClIN₄O₇
MW: 538,68 g/mol
MDL Number: MFCD03701153
CAS nummer: 5987-76-8

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 182.0 to 186.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR