Tantalum pentachloride, vattenfri ≥95.0%
Leverantör: TCI EUROPE
Fara
Synonyms: Tantalum (V) chloride
T2684-5G T2684-25G
TCIAT2684-5GEA 405 SEK
TCIAT2684-5G TCIAT2684-25G
Tantalum pentachloride, vattenfri ≥95.0%
Tantalum pentachloride
Formel: Cl₅Ta
MW: 358,21 g/mol
Kokpunkt: 239°C
Smältpunkt: 220°C
MDL Number: MFCD00011253
CAS nummer: 7721-01-9
EINECS: 231-755-6
UN: 3260
ADR: 8,II
Merck Index: 14,09054
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 95.0 %