L-Threonine benzyl ester hydrochloride ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE

Synonyms: (2S,3R)-Benzyl 2-amino-3-hydroxybutanoate hydrochloride, H-Thr-OBzl.HCl

T2677-1G T2677-5G
TCIAT2677-1GEA 415 SEK
TCIAT2677-1G TCIAT2677-5G
L-Threonine benzyl ester hydrochloride ≥98.0% (by HPLC, total nitrogen)
L-Threonine benzyl ester hydrochloride
Formel: C₁₁H₁₅NO₃
MW: 245,70 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00038977
CAS nummer: 33645-24-8

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 128.0 to 132.0 deg-C
Specific rotation [a]20/D -10.0 to -12.0 deg (C=1, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR